Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mblac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028372.1 | NP_082648.1 | 2 | 703 | ||
GenBank mRNA | AK046032.1 | - | 2 | 704 | ||
BC107206.1 | - | 2 | 423 | |||
BC107207.1 | - | 2 | 427 | |||
BC114401.1 | - | 2 | 424 |