Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278415.1 | NP_001265344.1 | 8 | 791 | ||
NM_028388.3 | NP_082664.1 | 7 | 694 | |||
GenBank mRNA | AK013511.1 | - | 7 | 645 | ||
AK078351.1 | - | 7 | 638 | |||
AK088193.1 | - | 4 | 433 | |||
AK146998.1 | - | 7 | 628 | |||
AK150404.1 | - | 8 | 728 | |||
AK151729.1 | - | 8 | 728 | |||
AK159460.1 | - | 7 | 611 | |||
AK166539.1 | - | 7 | 655 | |||
AK169143.1 | - | 7 | 665 | |||
AK185171.1 | - | 3 | 318 | |||
BC030946.1 | - | 7 | 623 | 5003524 | ||
CF106105.1 | - | 6 | 536 |