Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hepacam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175189.4 | NP_780398.2 | 2 | 306 | ||
GenBank mRNA | AK013637.1 | - | 2 | 307 | ||
AK081154.1 | - | 2 | 222 | |||
AK158265.1 | - | 2 | 544 | |||
BC082537.1 | - | 2 | 220 | |||
BC139057.1 | - | 2 | 296 | 8862079 | ||
BC139058.1 | - | 2 | 296 | 8862080 | ||
BY753058.1 | - | 2 | 541 |