Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Swi5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290552.1 | NP_001277481.1 | 3, 4 | 210 | ||
NM_175190.5 | NP_780399.1 | 3, 4 | 355 | |||
GenBank mRNA | AB597964.1 | - | 3, 4 | 203 | ||
AB597965.1 | - | 1, 2 | 107 | |||
AK013517.1 | - | 3, 4 | 315 | |||
AK031907.1 | - | 3, 4 | 323 | |||
AK041567.1 | - | 3, 4 | 297 | |||
AK088274.1 | - | 3, 4 | 316 | |||
AK166608.1 | - | 3, 4 | 299 | |||
AK216650.1 | - | 1, 2 | 124 | |||
BC021748.1 | - | 3, 4 | 305 | 3586619 |