Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnih3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160211.1 | NP_001153683.1 | 2 | 915 | ||
NM_001160212.1 | NP_001153684.1 | 2 | 915 | |||
NM_028408.3 | NP_082684.1 | 2 | 915 | |||
GenBank mRNA | AK013789.1 | - | 2 | 912 | ||
AK033911.1 | - | 2 | 913 | |||
BC115640.1 | - | 2 | 541 | 40103001 | ||
BC115641.1 | - | 2 | 541 | 40103002 | ||
BG800984.1 | - | 2 | 345 |