Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkrir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028410.1 | NP_082686.1 | 5 | 1892 | ||
GenBank mRNA | AK013663.1 | - | 5 | 1983 | ||
AK028175.1 | - | 1 | 997 | |||
AK217173.1 | - | 1 | 206 | |||
BC022684.1 | - | 1 | 107 | |||
BC049103.1 | - | 5 | 1892 | 6400126 |