Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2n2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028420.2 | NP_082696.1 | 2 | 275 | ||
GenBank mRNA | AK013788.1 | - | 2 | 222 | ||
BC147550.1 | - | 2 | 252 | 9056541 | ||
BC147736.2 | - | 2 | 252 | 9056685 | ||
BC147745.1 | - | 2 | 252 | 9056694 | ||
BC157896.1 | - | 2 | 252 | 9056547 | ||
CX215091.1 | - | 2 | 274 |