Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033371.2 | NP_203535.1 | 6 | 1480 | ||
GenBank mRNA | AK013817.1 | - | 2 | 228 | ||
AK134809.1 | - | 1 | 192 | |||
AK154865.1 | - | 6 | 1465 | |||
AK171452.1 | - | 6 | 1465 | |||
AK181600.1 | - | 1 | 24 | |||
AK220248.1 | - | 6 | 1480 | |||
AY010723.1 | - | 5 | 675 | |||
BC033318.1 | - | 5 | 1040 | |||
BC044775.1 | - | 4 | 1235 | |||
BC049958.1 | - | 4 | 1235 | |||
BC057450.1 | - | 5 | 1097 | 3966334 | ||
BC096620.1 | - | 6 | 1480 | 6390076 |