Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmpcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028431.2 | NP_082707.1 | 6 | 695 | ||
GenBank mRNA | AK013995.1 | - | 6 | 696 | ||
AK152673.1 | - | 6 | 696 | |||
AK169417.1 | - | 6 | 652 | |||
AK191207.1 | - | 2 | 155 | |||
BC038321.1 | - | 5 | 566 | |||
BC051216.1 | - | 5 | 502 | |||
BC060662.1 | - | 6 | 688 | |||
BC065095.1 | - | 6 | 688 | |||
BC087943.1 | - | 6 | 707 |