Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033167.3 | NP_001028339.1 | 2 | 1095 | ||
GenBank mRNA | AK147397.1 | - | 2 | 1096 |