Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam101a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028443.2 | NP_082719.1 | 1, 2 | 269 | ||
GenBank mRNA | AK014112.1 | - | 1, 2 | 299 | ||
BC053444.1 | - | 1, 2 | 269 | 6336076 |