Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wasl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167745.1 | NP_001161217.1 | 3, 4 | 688 | ||
NM_028459.2 | NP_082735.2 | 3, 4 | 688 | |||
GenBank mRNA | AJ318416.1 | - | 3, 4 | 406 | ||
AK014104.1 | - | 3, 4 | 682 | |||
AK140189.1 | - | 3, 4 | 606 | |||
AK142642.1 | - | 2, 3 | 164 | |||
AK143969.1 | - | 3, 4 | 669 | |||
AK144789.1 | - | 3, 4 | 664 | |||
AK147881.1 | - | 1, 2 | 75 | |||
AK159052.1 | - | 3, 4 | 640 | |||
BC019951.1 | - | 3, 4 | 578 | |||
BC055045.1 | - | 3, 4 | 664 | 6489610 | ||
BC058642.1 | - | 3, 4 | 688 |