Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddit4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030143.4 | NP_084419.2 | 3 | 755 | ||
GenBank mRNA | AF327512.1 | - | 2 | 510 | ||
AF335325.1 | - | 2 | 555 | |||
AK006617.1 | - | 3 | 754 | |||
AK018944.1 | - | 3 | 757 | |||
AK029362.1 | - | 1 | 323 | |||
AK053333.1 | - | 3 | 693 | |||
BC038131.1 | - | 3 | 654 | |||
BC147181.1 | - | 2 | 534 | 9056209 | ||
BC147182.1 | - | 2 | 534 | 9056220 |