Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038993.3 | NP_001034082.1 | 11 | 1459 | ||
NM_175201.5 | NP_780410.2 | 10 | 1489 | |||
GenBank mRNA | AK006888.1 | - | 9 | 1328 | ||
AK147608.1 | - | 11 | 1516 | |||
AK163346.1 | - | 6 | 620 | |||
BC060730.1 | - | 10 | 1466 | 6408623 | ||
BC062976.1 | - | 10 | 1403 | 6848803 |