Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166376.1 | NP_001159848.1 | 2, 3 | 347 | ||
NM_028595.4 | NP_082871.2 | 2, 3 | 347 | |||
GenBank mRNA | AF237910.1 | - | 2, 3 | 305 | ||
AK008652.1 | - | 2, 3 | 321 | |||
AK075852.1 | - | 2, 3 | 275 | |||
AK150397.1 | - | 2, 3 | 244 | |||
AK152258.1 | - | 2, 3 | 244 | |||
BC062247.1 | - | 2, 3 | 290 | 1550331 |