Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023712.3 | NP_076201.2 | 3, 4 | 1307 | ||
GenBank mRNA | AK154353.1 | - | 3, 4 | 797 | ||
AK166931.1 | - | 3, 4 | 799 | |||
AK202033.1 | - | 2, 3 | 209 | |||
BC002297.1 | - | 3, 4 | 326 | |||
BC026854.1 | - | 3, 4 | 768 | 4242049 | ||
BC085491.1 | - | 3, 4 | 751 | 6829415 |