Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163609.1 | NP_001157081.1 | 6 | 704 | ||
GenBank mRNA | AK010717.1 | - | 6 | 705 | ||
BC019368.1 | - | 3 | 170 | |||
BC116990.1 | - | 6 | 666 | 40126309 | ||
BC116992.1 | - | 6 | 666 | 40126311 | ||
BC144771.1 | - | 7 | 737 | 9053319 | ||
BC144772.1 | - | 5 | 546 | 9053320 |