Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111111.1 | NP_001104581.1 | 14 | 1453 | ||
GenBank mRNA | AB476647.1 | - | 13 | 1329 | ||
AB476648.1 | - | 14 | 1392 | |||
AK010773.1 | - | 13 | 1374 | |||
AK131109.1 | - | 1387 | ||||
AK155125.1 | - | 14 | 1443 | |||
AK157410.1 | - | 14 | 1432 | |||
AK158186.1 | - | 13 | 1567 | |||
BC060054.1 | - | 13 | 1954 |