Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166173.1 | NP_001159645.1 | 17 | 1637 | ||
NM_001166174.1 | NP_001159646.1 | 15 | 1544 | |||
NM_028618.2 | NP_082894.1 | 4 | 228 | |||
NM_172899.4 | NP_766487.3 | 16 | 1589 | |||
GenBank mRNA | AK003695.1 | - | 4 | 231 | ||
AK081753.1 | - | 17 | 1639 | |||
AY444557.1 | - | 15 | 1501 | |||
AY444558.1 | - | 4 | 231 | |||
AY622962.1 | - | 4 | 209 | |||
AY622963.1 | - | 15 | 1448 | |||
BC064724.1 | - | 16 | 1582 | 30435974 |