Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166173.1 | NP_001159645.1 | 17, 18 | 1675 | ||
NM_001166174.1 | NP_001159646.1 | 15, 16 | 1582 | |||
NM_028618.2 | NP_082894.1 | 4, 5 | 266 | |||
NM_172899.4 | NP_766487.3 | 16, 17 | 1627 | |||
GenBank mRNA | AK003695.1 | - | 4, 5 | 269 | ||
AK081753.1 | - | 17, 18 | 1677 | |||
AY444557.1 | - | 15, 16 | 1539 | |||
AY444558.1 | - | 4, 5 | 269 | |||
AY622962.1 | - | 4, 5 | 247 | |||
AY622963.1 | - | 15, 16 | 1486 | |||
BC064724.1 | - | 16, 17 | 1620 | 30435974 |