Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Whrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008791.2 | NP_001008791.1 | 13 | 3380 | ||
NM_001008792.2 | NP_001008792.1 | 12 | 3359 | |||
NM_001008793.2 | NP_001008793.1 | 12 | 3344 | |||
NM_028640.2 | NP_082916.1 | 12 | 3347 | |||
GenBank mRNA | AK004110.1 | - | 2 | 77 | ||
AK122523.1 | - | 12 | 2475 | |||
AY739114.1 | - | 12 | 3307 | |||
AY739115.1 | - | 13 | 3340 | |||
AY739116.1 | - | 12 | 3319 | |||
AY739117.1 | - | 12 | 3304 | |||
AY739118.1 | - | 9 | 1956 | |||
AY739119.1 | - | 8 | 1923 | |||
AY739120.1 | - | 7 | 1710 | |||
AY739121.1 | - | 7 | 1828 | |||
AY739122.1 | - | 8 | 2160 | |||
BC150757.1 | - | 12 | 2704 | 9087668 |