Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163551.1 | NP_001157023.1 | 5 | 433 | ||
NM_028648.3 | NP_082924.2 | 4 | 409 | |||
GenBank mRNA | AK015291.1 | - | 4 | 410 | ||
BC049645.1 | - | 5 | 443 | |||
BC103787.1 | - | 5 | 444 | 30919261 | ||
BY714849.1 | - | 4 | 410 |