Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290662.1 | NP_001277591.1 | 10 | 1003 | ||
NM_134471.4 | NP_608301.3 | 10 | 1044 | |||
GenBank mRNA | AK088047.1 | - | 10 | 985 | ||
AK150222.1 | - | 10 | 975 | |||
AK161309.1 | - | 10 | 1003 | |||
BC006841.1 | - | 10 | 1004 | 3598157 |