Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028665.4 | NP_082941.2 | 2 | 1133 | ||
GenBank mRNA | AK029511.1 | - | 2 | 1135 | ||
AK029888.1 | - | 2 | 1094 | |||
AK033599.1 | - | 2 | 1122 | |||
AK048925.1 | - | 2 | 1027 | |||
AK133339.1 | - | 1094 | ||||
BC030885.1 | - | 2 | 762 | |||
BC100433.1 | - | 2 | 1127 | |||
BC145456.1 | - | 2 | 251 | 9054017 | ||
BC145457.1 | - | 2 | 251 | |||
BC145458.1 | - | 2 | 251 |