Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100505025 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133139.1 | - | 1 | 226 |
Gene Symbol: 4930429H19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015238.1 | - | 76 |