Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176837.2 | NP_789807.1 | 11 | 1605 | ||
GenBank mRNA | AK014728.1 | - | 11 | 1553 | ||
AK078522.1 | - | 6 | 657 | |||
AK162202.1 | - | 7 | 848 | |||
BC030858.1 | - | 11 | 1605 | 4194996 |