Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn34c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281537.1 | NP_001268466.1 | 2 | 630 | ||
GenBank mRNA | AK015117.1 | - | 2 | 631 |