Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025947.2 | NP_001021118.1 | 15 | 1924 | ||
NM_001276340.1 | NP_001263269.1 | 16 | 1975 | |||
NM_001276341.1 | NP_001263270.1 | 15 | 1719 | |||
NM_152816.3 | NP_690029.2 | 16 | 1963 | |||
GenBank mRNA | AB079133.1 | - | 16 | 1809 | ||
AK018195.1 | - | 16 | 1903 | |||
AK051443.1 | - | 15 | 1853 | |||
AK080871.1 | - | 15 | 1844 | |||
AK135420.1 | - | 1 | 831 | |||
AK154123.1 | - | 17 | 1804 | |||
BC040777.1 | - | 15 | 1689 | 4948368 | ||
BC079635.1 | - | 15 | 1773 | 6400313 | ||
BC085158.1 | - | 16 | 1868 | |||
BE633575.1 | - | 3 | 245 | |||
CK781757.1 | - | 1 | 89 |