Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079814.1 | NP_001073282.1 | 12 | 1134 | ||
NM_028720.2 | NP_082996.2 | 12 | 1116 | |||
GenBank mRNA | AK014456.1 | - | 12 | 1056 | ||
AK040841.1 | - | 12 | 1066 | |||
AK145277.1 | - | 12 | 1067 | |||
AK150349.1 | - | 12 | 1060 | |||
AK151832.1 | - | 12 | 1082 | |||
AK152586.1 | - | 12 | 1082 | |||
AK152887.1 | - | 12 | 1060 | |||
BC006893.1 | - | 12 | 1050 | 3599477 | ||
CF174076.1 | - | 7 | 472 |