Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028721.3 | NP_082997.3 | 6 | 1230 | ||
NM_172460.1 | NP_766048.1 | 6 | 1230 | |||
GenBank mRNA | AK048877.1 | - | 6 | 1003 | ||
AK052281.1 | - | 6 | 1017 | |||
AK133217.1 | - | 6 | 1231 | |||
AK173327.1 | - | 6 | 809 | |||
AY259499.1 | - | 6 | 1170 | |||
BC115724.1 | - | 6 | 760 | 40086844 |