Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177615.1 | NP_001171086.1 | 2 | 391 | ||
NM_001177616.1 | NP_001171087.1 | 3 | 398 | |||
NM_001177617.1 | NP_001171088.1 | 3 | 565 | |||
NM_001177618.1 | NP_001171089.1 | 2 | 96 | |||
NM_001177619.1 | NP_001171090.1 | 3 | 763 | |||
NM_001177620.1 | NP_001171091.1 | 4 | 338 | |||
NM_001177623.1 | NP_001171094.1 | 3 | 327 | |||
NM_028755.3 | NP_083031.1 | 3 | 300 | |||
NM_033264.2 | NP_150289.1 | 2 | 231 | |||
GenBank mRNA | AF324451.1 | - | 2 | 191 | ||
AK002950.2 | - | 3 | 284 | |||
AK029467.1 | - | 3 | 764 | |||
AK032417.1 | - | 3 | 324 | |||
AK032543.1 | - | 11 | ||||
AK045507.1 | - | 3 | 765 | |||
AK050613.1 | - | 3 | 328 | |||
AK137974.1 | - | 3 | 566 | |||
AK153892.1 | - | 4 | 1046 | |||
AK159004.1 | - | 2 | 370 | |||
AK160860.1 | - | 4 | 336 | |||
BC027107.1 | - | 3 | 288 | |||
BC053001.1 | - | 1 | 62 | 5686446 | ||
BC098368.1 | - | 3 | 345 | |||
BC128376.1 | - | 2 | 112 | |||
BY126803.1 | - | 3 | 300 | |||
BY271626.1 | - | 2 | 396 | |||
BY275281.1 | - | 3 | 399 | |||
BY290692.1 | - | 2 | 97 |