Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177615.1 | NP_001171086.1 | 4 | 586 | ||
NM_001177616.1 | NP_001171087.1 | 5 | 593 | |||
NM_001177617.1 | NP_001171088.1 | 5 | 760 | |||
NM_001177618.1 | NP_001171089.1 | 4 | 291 | |||
NM_001177619.1 | NP_001171090.1 | 5 | 958 | |||
NM_001177620.1 | NP_001171091.1 | 6 | 533 | |||
NM_001177623.1 | NP_001171094.1 | 5 | 522 | |||
NM_028755.3 | NP_083031.1 | 5 | 495 | |||
NM_033264.2 | NP_150289.1 | 4 | 426 | |||
GenBank mRNA | AF324451.1 | - | 4 | 386 | ||
AK002950.2 | - | 5 | 479 | |||
AK029467.1 | - | 5 | 959 | |||
AK032417.1 | - | 5 | 519 | |||
AK032543.1 | - | 3 | 206 | |||
AK034376.1 | - | 3 | 139 | |||
AK045507.1 | - | 5 | 960 | |||
AK050613.1 | - | 5 | 523 | |||
AK137974.1 | - | 5 | 761 | |||
AK153892.1 | - | 6 | 1241 | |||
AK159004.1 | - | 4 | 565 | |||
AK160860.1 | - | 6 | 531 | |||
BC027107.1 | - | 5 | 483 | |||
BC053001.1 | - | 3 | 257 | 5686446 | ||
BC098368.1 | - | 5 | 540 | |||
BC128376.1 | - | 4 | 307 |