Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028787.4 | NP_083063.2 | 11 | 1298 | ||
GenBank mRNA | AK004892.2 | - | 11 | 1279 | ||
AK076339.1 | - | 11 | 1292 | |||
AK145848.1 | - | 11 | 1299 | |||
AK169012.1 | - | 11 | 1160 | |||
AK171951.1 | - | 11 | 1272 | |||
BC026858.1 | - | 11 | 1157 | 4014064 |