Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023738.4 | NP_076227.1 | 3 | 390 | ||
GenBank mRNA | AK004894.2 | - | 3 | 391 | ||
AK054410.1 | - | 3 | 397 | |||
AK089235.1 | - | 3 | 394 | |||
AK089256.1 | - | 2 | 465 | |||
AK129009.1 | - | 3 | 302 | |||
AK170421.1 | - | 3 | 383 | |||
AK170644.1 | - | 3 | 389 | |||
AK172467.1 | - | 3 | 352 | |||
BC037693.1 | - | 1 | 109 | |||
BC138843.1 | - | 3 | 307 | 8861865 |