Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028791.5 | NP_083067.1 | 13 | 1648 | ||
GenBank mRNA | AK005044.2 | - | 13 | 1555 | ||
AK042060.1 | - | 13 | 1594 | |||
AK043733.1 | - | 13 | 1585 | |||
AK049190.1 | - | 13 | 1632 | |||
AK080379.1 | - | 13 | 1649 | |||
AK088624.1 | - | 13 | 1583 | |||
AK150392.1 | - | 13 | 1574 | |||
AK151194.1 | - | 13 | 1574 | |||
AK152276.1 | - | 13 | 1574 | |||
AK152452.1 | - | 13 | 1555 | |||
AK152799.1 | - | 13 | 1574 | |||
AK153021.1 | - | 13 | 1574 | |||
AK153066.1 | - | 13 | 1574 | |||
AK154773.1 | - | 13 | 1620 | |||
AK172883.1 | - | 10 | 1131 | |||
AK183115.1 | - | 1 | 152 | |||
AK196080.1 | - | 1 | 152 | |||
BC024691.1 | - | 13 | 1570 | 3668655 |