Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028791.5 | NP_083067.1 | 19, 20 | 2214 | ||
GenBank mRNA | AK005044.2 | - | 19, 20 | 2121 | ||
AK042060.1 | - | 19, 20 | 2160 | |||
AK043733.1 | - | 19, 20 | 2151 | |||
AK049190.1 | - | 19, 20 | 2198 | |||
AK080379.1 | - | 19, 20 | 2215 | |||
AK088624.1 | - | 19, 20 | 2149 | |||
AK150392.1 | - | 19, 20 | 2140 | |||
AK151194.1 | - | 19, 20 | 2140 | |||
AK152276.1 | - | 19, 20 | 2140 | |||
AK152452.1 | - | 19, 20 | 2121 | |||
AK152799.1 | - | 19, 20 | 2140 | |||
AK153021.1 | - | 19, 20 | 2140 | |||
AK153066.1 | - | 19, 20 | 2140 | |||
AK154773.1 | - | 19, 20 | 2186 | |||
AK172883.1 | - | 16, 17 | 1696 | |||
BC024691.1 | - | 19, 20 | 2136 | 3668655 |