Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028791.5 | NP_083067.1 | 5, 6 | 720 | ||
GenBank mRNA | AK005044.2 | - | 5, 6 | 627 | ||
AK042060.1 | - | 5, 6 | 666 | |||
AK043733.1 | - | 5, 6 | 657 | |||
AK049190.1 | - | 5, 6 | 704 | |||
AK080379.1 | - | 5, 6 | 721 | |||
AK088624.1 | - | 5, 6 | 655 | |||
AK150392.1 | - | 5, 6 | 646 | |||
AK151194.1 | - | 5, 6 | 646 | |||
AK152276.1 | - | 5, 6 | 646 | |||
AK152452.1 | - | 5, 6 | 627 | |||
AK152799.1 | - | 5, 6 | 646 | |||
AK153021.1 | - | 5, 6 | 646 | |||
AK153066.1 | - | 5, 6 | 646 | |||
AK154773.1 | - | 5, 6 | 692 | |||
AK172883.1 | - | 2, 3 | 203 | |||
AK182191.1 | - | 1, 2 | 108 | |||
AK186244.1 | - | 1, 2 | 108 | |||
AK212192.1 | - | 1, 2 | 108 | |||
BC024691.1 | - | 5, 6 | 642 | 3668655 |