Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023740.2 | NP_076229.2 | 4 | 695 | ||
GenBank mRNA | AF176814.1 | - | 4 | 668 | ||
AK005255.1 | - | 2 | 291 | |||
AK151505.1 | - | 4 | 677 | |||
AK168042.1 | - | 4 | 670 | |||
AK183281.1 | - | 1 | 102 | |||
AK186864.1 | - | 1 | 78 | |||
AK188228.1 | - | 1 | 78 | |||
AK202579.1 | - | 1 | 68 | |||
AK205856.1 | - | 1 | 78 | |||
AK211692.1 | - | 1 | 78 | |||
AK214551.1 | - | 1 | 78 | |||
AK218025.1 | - | 1 | 78 | |||
AK219188.1 | - | 1 | 89 | |||
BC010835.1 | - | 4 | 659 | 4239913 |