Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028797.1 | NP_083073.1 | 4, 5 | 343 | ||
GenBank mRNA | AK005675.1 | - | 4, 5 | 321 | ||
BU945975.1 | - | 4, 5 | 345 | |||
CO812705.1 | - | 4, 5 | 367 |