Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028809.1 | NP_083085.1 | 2 | 243 | ||
GenBank mRNA | AK008255.1 | - | 2 | 243 | ||
AK037305.1 | - | 1 | 3134 | |||
BC024482.1 | - | 2 | 266 | 4977391 |