Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700016H13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163550.1 | NP_001157022.1 | 4 | 539 | ||
NM_028824.1 | NP_083100.1 | 4 | 690 | |||
GenBank mRNA | AK006033.1 | - | 4 | 692 | ||
BC049644.1 | - | 4 | 682 | 6705673 |