Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001565.2 | NP_001001565.1 | 5 | 2533 | ||
NM_001001566.3 | NP_001001566.1 | 4 | 2390 | |||
GenBank mRNA | AK006466.1 | - | 4 | 1761 | ||
AK155424.1 | - | 4 | 2051 | |||
AK160734.1 | - | 3 | 2638 | |||
BC030889.1 | - | 4 | 1878 | |||
BC057050.1 | - | 5 | 2369 | |||
BC060598.1 | - | 5 | 2369 | |||
BC071273.1 | - | 4 | 2190 | 6334418 |