Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175207.4 | NP_780416.2 | 3 | 1327 | ||
GenBank mRNA | AK010867.1 | - | 3 | 1490 | ||
AK032641.1 | - | 3 | 1328 | |||
AK051108.1 | - | 3 | 1347 | |||
AK172167.1 | - | 4 | 1409 | |||
BC023687.1 | - | 3 | 1301 | 5343549 |