Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133756.4 | NP_598517.1 | 3 | 234 | ||
GenBank mRNA | AK010393.1 | - | 3 | 248 | ||
AK077645.1 | - | 3 | 235 | |||
AK145957.1 | - | 231 | ||||
AK146686.1 | - | 246 | ||||
AK163635.1 | - | 3 | 711 | |||
BC020174.1 | - | 3 | 234 | 4505118 | ||
BC064761.1 | - | 3 | 242 | |||
BC096466.1 | - | 3 | 227 | 3711417 |