Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128170.2 | NP_001121642.1 | 16 | 2645 | ||
NM_001128171.2 | NP_001121643.1 | 16 | 2770 | |||
NM_001276279.1 | NP_001263208.1 | 14 | 2221 | |||
NM_173369.3 | NP_775545.1 | 15 | 2636 | |||
GenBank mRNA | AK122389.1 | - | 16 | 2751 | ||
AK133479.1 | - | 15 | 2600 | |||
AK134926.1 | - | 3 | 328 | |||
AK147204.1 | - | 16 | 2613 | |||
AK147426.1 | - | 16 | 2608 | |||
AK167424.1 | - | 14 | 2185 | |||
BC024596.1 | - | 3 | 314 | |||
BC028885.1 | - | 8 | 2291 | |||
BC042438.1 | - | 15 | 2589 | 3983771 | ||
BC049879.1 | - | 6 | 498 |