Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aven | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165935.1 | NP_001159407.1 | 4 | 421 | ||
NM_028844.3 | NP_083120.2 | 5 | 751 | |||
GenBank mRNA | AK005934.1 | - | 4 | 476 | ||
AK006808.1 | - | 5 | 752 | |||
AK046479.1 | - | 677 | ||||
BC100725.1 | - | 6 | 614 | 40045263 | ||
BC100735.1 | - | 6 | 628 | |||
BX528932.1 | - | 3 | 281 | |||
DV646415.1 | - | 4 | 423 |