Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028863.1 | NP_083139.1 | 4 | 350 | ||
GenBank mRNA | AK002936.1 | - | 4 | 349 | ||
AK019988.1 | - | 3 | 517 | |||
AK210710.1 | - | 3 | 169 | |||
BC022589.1 | - | 2 | 1083 | 4037176 | ||
BY125913.1 | - | 3 | 228 |