Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028873.4 | NP_083149.3 | 7 | 2173 | ||
GenBank mRNA | AK017830.1 | - | 7 | 2187 | ||
AK032681.1 | - | 7 | 2155 | |||
AK045345.1 | - | 7 | 2231 | |||
AK045445.1 | - | 7 | 2267 | |||
AK082579.1 | - | 7 | 2174 | |||
AK083878.1 | - | 7 | 2184 | |||
AK159388.1 | - | 7 | 2186 | |||
AK168935.1 | - | 7 | 2265 | |||
BC011146.1 | - | 7 | 2238 | 4016209 |