Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrguk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028886.1 | NP_083162.1 | 1, 2 | 314 | ||
GenBank mRNA | AK014976.1 | - | 1, 2 | 318 | ||
AK029666.1 | - | 1, 2 | 313 | |||
AK045903.1 | - | 1, 2 | 314 |