Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rptor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB082952.1 | - | 1 | 18 | ||
AK029341.1 | - | 1 | 813 | |||
AK087217.1 | - | 1 | 834 | |||
AK144082.1 | - | 1 | 813 | |||
AK161011.1 | - | 1 | 924 | |||
AK172276.1 | - | 1 | 811 | |||
BC060140.1 | - | 1 | 130 | |||
BC118971.1 | - | 1 | 13 | |||
BC148309.2 | - | 1 | 18 |